Merck
  • Phosphorylation of Ser283 enhances the stiffness of the tropomyosin head-to-tail overlap domain.
Phosphorylation of Ser283 enhances the stiffness of the tropomyosin head-to-tail overlap domain.
Archives of biochemistry and biophysics (2015-03-03)
William Lehman, Greg Medlock, Xiaochuan Edward Li, Worawit Suphamungmee, An-Yue Tu, Anja Schmidtmann, Zoltán Ujfalusi, Stefan Fischer, Jeffrey R Moore, Michael A Geeves, Michael Regnier
ABSTRACT
The ends of coiled-coil tropomyosin molecules are joined together by nine to ten residue-long head-to-tail "overlapping domains". These short four-chained interconnections ensure formation of continuous tropomyosin cables that wrap around actin filaments. Molecular Dynamics simulations indicate that the curvature and bending flexibility at the overlap is 10-20% greater than over the rest of the molecule, which might affect head-to-tail filament assembly on F-actin. Since the penultimate residue of striated muscle tropomyosin, Ser283, is a natural target of phosphorylating enzymes, we have assessed here if phosphorylation adjusts the mechanical properties of the tropomyosin overlap domain. MD simulations show that phosphorylation straightens the overlap to match the curvature of the remainder of tropomyosin while stiffening it to equal or exceed the rigidity of canonical coiled-coil regions. Corresponding EM data on phosphomimetic tropomyosin S283D corroborate these findings. The phosphorylation-induced change in mechanical properties of tropomyosin likely results from electrostatic interactions between C-terminal phosphoSer283 and N-terminal Lys12 in the four-chain overlap bundle, while promoting stronger interactions among surrounding residues and thus facilitating tropomyosin cable assembly. The stiffening effect of D283-tropomyosin noted correlates with previously observed enhanced actin-tropomyosin activation of myosin S1-ATPase, suggesting a role for the tropomyosin phosphorylation in potentiating muscle contraction.
Product Description
Serine, European Pharmacopoeia (EP) Reference Standard
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)