Merck
  • Heterocyclic ring extension of androstenedione: synthesis and cytotoxicity of fused pyran, pyrimidine and thiazole derivatives.
Heterocyclic ring extension of androstenedione: synthesis and cytotoxicity of fused pyran, pyrimidine and thiazole derivatives.
Steroids (2014-05-06)
Rafat M Mohareb, Nermeen S Abbas, Mahmoud A Abdelaziz
ABSTRACT
The reaction of androstenedione with either malononitrile or ethyl cyanoacetate and aromatic aldehydes 2a-c gave the pyran derivatives 4a-f, respectively. On the other hand, the reaction of androstenedione with thiourea and the aromatic aldehydes 2a-c gave the pyrimidine derivatives 6a-c, respectively. Compound 6b reacted with 2-bromo-1-arylethanone derivatives 7a-d to give the indeno[2,1-e]thiazole derivatives 8a-d. Some of the produced compounds were used for further heterocyclization reactions. The cytotoxicity of the newly obtained products was evaluated against some cancer cell lines and a normal cell line.
Product Description
Supelco
4-Androstene-3,17-dione, VETRANAL®, analytical standard
SAFC
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source