Merck
  • Alternative reagents for methotrexate as immobilizing anchor moieties in the optimization of MASPIT: synthesis and biological evaluation.
Alternative reagents for methotrexate as immobilizing anchor moieties in the optimization of MASPIT: synthesis and biological evaluation.
Chembiochem : a European journal of chemical biology (2015-02-18)
Dries J H De Clercq, Martijn D P Risseeuw, Izet Karalic, Anne-Sophie De Smet, Dieter Defever, Jan Tavernier, Sam Lievens, Serge Van Calenbergh
ABSTRACT
We report the evaluation of two alternative chemical dimerizer approaches aimed at increasing the sensitivity of MASPIT, a three-hybrid system that enables small-molecule target protein profiling in intact human cells. To circumvent the potential limitations related to the binding of methotrexate (MTX) to endogenous human dihydrofolate reductase (DHFR), we explored trimethoprim (TMP) as an alternative prokaryote-specific DHFR ligand. MASPIT evaluation of TMP fusion compounds with tamoxifen, reversine, and simvastatin as model baits, resulted in dose-response curves shifted towards lower EC50 values than those of their MTX congeners. Furthermore, a scalable azido-TMP reagent was synthesized that displayed a similar improvement in sensitivity, possibly owing to increased membrane permeability relative to the MTX anchor. Applying the SNAP-tag approach to introduce a covalent bond into the system, on the other hand, produced an inferior readout than in the MTX- or TMP-tag based assay.
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, ACS reagent
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Deionized
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, HPLC Plus
Supelco
Water, suitable for ion chromatography
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%