Merck
  • Direct thiophenylation accompanying orthoester-cleavage of 1,2,4-O-orthoacetyl-3,6-O-(o-xylylene)glucopyranose.
Direct thiophenylation accompanying orthoester-cleavage of 1,2,4-O-orthoacetyl-3,6-O-(o-xylylene)glucopyranose.
Carbohydrate research (2014-12-17)
Takuya Uchino, Yusuke Tomabechi, Atsushi Fukumoto, Hidetoshi Yamada
ABSTRACT
The 3,6-O-(o-xylylene) bridge locks the conformation of glucopyranose to an axial-rich form. Although the conformational lock induces complete β-selectivity in a glycosylation reaction, the leaving group of the glycosyl donor is limited to fluorine. On the other hand, the bridge confers the furanose-preferred property to glucose, which makes synthesis of corresponding pyranosyl derivatives that equip various leaving groups difficult. This problem was solved through direct phenylthio glucosidation of 3,6-O-(o-xylylene)-1,2,4-O-orthoacetylglucose accompanying cleavage of the orthoester moiety. This paper describes the process of establishing direct thiophenylation. This process reduced the synthetic steps for the known glucopyranosyl fluoride and will expand application of conformationally locked glycosyl donors.
Product Description
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Chloroform, analytical standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%