Merck
  • Screening for impact of popular herbs improving mental abilities on the transcriptional level of brain transporters.
Screening for impact of popular herbs improving mental abilities on the transcriptional level of brain transporters.
Acta pharmaceutica (Zagreb, Croatia) (2014-06-11)
Przemyslaw M Mrozikiewicz, Anna Bogacz, Joanna Bartkowiak-Wieczorek, Radoslaw Kujawski, Przemyslaw L Mikolajczak, Marcin Ozarowski, Boguslaw Czerny, Beata Mrozikiewicz-Rakowska, Edmund Grzeskowiak
ABSTRACT
There are a number of compounds that can modify the activity of ABC (ATP-binding cassette) and SLC (solute carrier) transporters in the blood-brain barrier (BBB). The aim of this study was to investigate the effect of natural and synthetic substances on the expression level of genes encoding transporters present in the BBB (mdr1a, mdr1b, mrp1, mrp2, oatp1a4, oatp1a5 and oatp1c1). Our results showed that verapamil caused the greatest reduction in the mRNA level while other synthetic (piracetam, phenobarbital) and natural (codeine, cyclosporine A, quercetin) substances showed a selective inhibitory effect. Further, the extract from the roots of Panax ginseng C. A. Meyer exhibited a decrease of transcription against selected transporters whereas the extract from Ginkgo biloba L. leaves resulted in an increase of the expression level of tested genes, except for mrp2. Extract from the aerial parts of Hypericum perforatum L. was the only one to cause an increased mRNA level for mdr1 and oatp1c1. These findings suggest that herbs can play an important role in overcoming the BBB and multidrug resistance to pharmacotherapy of brain cancer and mental disorders, based on the activity of selected drug-metabolizing enzymes and transporters located in the BBB.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Codeine solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Codeine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(±)-CPP, solid
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Ciclosporin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Piracetam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Piracetam
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%