Merck
  • Inactivation of a GAL4-like transcription factor improves cell fitness and product yield in glycoengineered Pichia pastoris strains.
Inactivation of a GAL4-like transcription factor improves cell fitness and product yield in glycoengineered Pichia pastoris strains.
Applied and environmental microbiology (2014-10-26)
Bo Jiang, Rebecca Argyros, John Bukowski, Stephanie Nelson, Nathan Sharkey, Sehoon Kim, Victoria Copeland, Robert C Davidson, Ronghua Chen, Jun Zhuang, Natarajan Sethuraman, Terrance A Stadheim
ABSTRACT
With a completely reengineered and humanized glycosylation pathway, glycoengineered Pichia pastoris has emerged as a promising production host for the manufacture of therapeutic glycoproteins. However, the extensive genetic modifications have also negatively affected the overall fitness levels of the glycoengineered host cells. To make glycoengineered Pichia strains more compatible with a scalable industrial fermentation process, we sought to identify genetic solutions to broadly improve cell robustness during fermentation. In this study, we report that mutations within the Pichia pastoris ATT1 (PpATT1) gene (a homolog of the Saccharomyces cerevisiae GAL4 [ScGAL4] transcriptional activator) dramatically increased the cellular fitness levels of glycoengineered Pichia strains. We demonstrate that deletion of the PpATT1 gene enabled glycoengineered Pichia strains to improve their thermal tolerance levels, reduce their cell lysis defects, and greatly improve fermentation robustness. The extension of the duration of fermentation enabled the PpATT1-modified glycoengineered Pichia strains to increase their product yields significantly without any sacrifice in product quality. Because the ATT1 gene could be deleted from any Pichia strains, including empty hosts and protein-expressing production strains alike, we suggest that the findings described in this study are broadly applicable to any Pichia strains used for the production of therapeutic proteins, including monoclonal antibodies, Fc fusions, peptides, hormones, and growth factors.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material