Merck
  • Protein kinase C regulates vascular calcification via cytoskeleton reorganization and osteogenic signaling.
Protein kinase C regulates vascular calcification via cytoskeleton reorganization and osteogenic signaling.
Biochemical and biophysical research communications (2014-12-03)
Kyunghee Lee, Hyunsoo Kim, Daewon Jeong
ABSTRACT
Vascular calcification is an active cell-mediated process that reduces elasticity of blood vessels and increases blood pressure. Until now, the molecular basis of vascular calcification has not been fully understood. We previously reported that microtubule disturbances mediate vascular calcification. Here, we found that protein kinase C (PKC) signaling acted as a novel coordinator between cytoskeletal changes and hyperphosphatemia-induced vascular calcification. Phosphorylation and expression of both PKCα and PKCδ decreased during inorganic phosphate (Pi)-induced vascular smooth muscle cell (VSMC) calcification. Knockdown of PKC isoforms by short interfering RNA as well as PKC inactivation by Go6976 or rottlerin treatment revealed that specific inhibition of PKCα and PKCδ accelerated Pi-induced calcification both in VSMCs and ex vivo aorta culture through upregulation of osteogenic signaling. Additionally, inhibition of PKCα and PKCδ induced disassembly of microtubule and actin, respectively. In summary, our results indicate that cytoskeleton perturbation via PKCα and PKCδ inactivation potentiates vascular calcification through osteogenic signal induction.
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M