Merck
  • Effects of naproxen on cell proliferation and genotoxicity in MG-63 osteosarcoma cell line.
Effects of naproxen on cell proliferation and genotoxicity in MG-63 osteosarcoma cell line.
Journal of toxicology and environmental health. Part A (2014-07-30)
Isabel Correia, Regina Arantes-Rodrigues, Rosário Pinto-Leite, Isabel Gaivão
ABSTRACT
The purpose of this study was to determine the efficacy of naproxen, a nonsteroidal anti-inflammatory drug, on the MG-63 human osteosarcoma cell line. MG-63 cells were exposed to naproxen in a wide range of concentrations of 0.03, 0.05, 0.1, 0.42, 0.83, and 1.67 mg/ml for 72 h. The activity of naproxen was assessed by the following assays: cell morphology; 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl tetrazolium bromide (MTT) method; terminal deoxynucleotidyl transferase-mediated dUTP nick end labeling (TUNEL) assay; comet assay; and acridine orange and monodansylcadaverine (MDC) staining. Naproxen exerted a significant inhibitory effect on MG-63 cell proliferation, in a concentration-dependent manner, in all treatment groups compared with untreated cells. An increase in frequency of DNA damage, apoptotic bodies, apoptotic cells, and autophagic vacuoles was observed in MG-63-treated cells. Although future studies are needed, these findings suggest that naproxen may lead to improvements in treatment of patients with osteosarcoma.
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Naproxen, meets USP testing specifications
SAFC
L-Glutamine
Naproxen, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Naproxen, VETRANAL®, analytical standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture