Merck
  • Gene expression levels of Casein kinase 1 (CK1) isoforms are correlated to adiponectin levels in adipose tissue of morbid obese patients and site-specific phosphorylation mediated by CK1 influences multimerization of adiponectin.
Gene expression levels of Casein kinase 1 (CK1) isoforms are correlated to adiponectin levels in adipose tissue of morbid obese patients and site-specific phosphorylation mediated by CK1 influences multimerization of adiponectin.
Molecular and cellular endocrinology (2015-03-01)
Pengfei Xu, Pamela Fischer-Posovszky, Joachim Bischof, Peter Radermacher, Martin Wabitsch, Doris Henne-Bruns, Anna-Maria Wolf, Andreas Hillenbrand, Uwe Knippschild
ABSTRACT
White adipose tissue has now been recognized as an important endocrine organ secreting bioactive molecules termed adipocytokines. In obesity, anti-inflammatory adipocytokines like adiponectin are decreased while pro-inflammatory factors are over-produced. These changes contribute to the development of insulin resistance and obesity-associated diseases. Since members of the casein kinase 1 (CK1) family are involved in the regulation of various signaling pathways we ask here whether they are able to modulate the functions of adiponectin. We show that CK1δ and ε are expressed in adipose tissue and that the expression of CK1 isoforms correlates with that of adiponectin. Furthermore, adiponectin co-immunoprecipitates with CK1δ and CK1ε and is phosphorylated by CK1δ at serine 174 and threonine 235, thereby influencing the formation of adiponectin oligomeric complexes. Furthermore, inhibition of CK1δ in human adipocytes by IC261 leads to an increase in basal and insulin-stimulated glucose uptake. In summary, our data indicate that site-specific phosphorylation of adiponectin, especially at sites targeted by CK1δ in vitro, provides an additional regulatory mechanism for modulating adiponectin complex formation and function.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Akt1 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥75% (SDS-PAGE)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG