Merck
  • Influenza viral load and peramivir kinetics after single administration and proposal of regimens for peramivir administration against resistant variants.
Influenza viral load and peramivir kinetics after single administration and proposal of regimens for peramivir administration against resistant variants.
Antimicrobial agents and chemotherapy (2014-12-31)
Masatoki Sato, Masaki Ito, Shigeo Suzuki, Hiroko Sakuma, Aya Takeyama, Shinichi Oda, Masahiro Watanabe, Koichi Hashimoto, Kyohei Miyazaki, Yukihiko Kawasaki, Mitsuaki Hosoya
ABSTRACT
We estimated the efficacy of the current single administration of peramivir on the basis of peramivir pharmacokinetics in the upper respiratory tract (URT) and determined the predictive peramivir concentration-time curve to assess its efficacy against viruses with decreased susceptibility to neuraminidase inhibitors. Serum, nasal swab, or aspiration samples were collected from 28 patients treated with 10 mg/kg body weight peramivir. The sequential influenza viral RNA load and susceptibility after peramivir administration were measured using a quantitative real-time reverse transcription-PCR and neuraminidase inhibition assay. The peramivir concentrations in the serum and URT after a single administration at 10 mg/kg were measured, and the predictive blood and URT peramivir concentration-time curves were determined to assess various administration regimens against resistant variants. The peramivir concentration decreased to <0.1% of the maximum concentration of drug in serum (Cmax) at 24 h after administration. Rapid elimination of peramivir from the URT by 48 h after administration may contribute to an increase in the influenza A viral load after day 3 but not to a decrease in the influenza B viral load, despite the absence of a decrease in the susceptibility to peramivir. A longer maintenance of a high level of peramivir in the URT is expected by divided administration rather than once-daily administration. When no clinical improvement is observed in patients with normal susceptibility influenza A and B, peramivir readministration should be considered. In severe cases caused by resistant variants, better inhibitory effectiveness and less frequent adverse events are expected by divided administration rather than once-daily administration with an increased dosage.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%