Merck
Role of zinc trafficking in male fertility: from germ to sperm.
Human reproduction (Oxford, England) (2014-04-29)
Carlo Foresta, Andrea Garolla, Ilaria Cosci, Massimo Menegazzo, Marco Ferigo, Valentina Gandin, Luca De Toni
ABSTRACT
What are the dynamics of zinc (Zn) trafficking in sperm, at the testicular, epididymal and ejaculate levels? Zn transporters are peculiarly expressed in the cells of the germ line and Zn uptake is maximal at the post-epididymal phase, where Zn is involved in the regulation of sperm functions. Zn is known to influence several phases of sperm life, from germ cell development to spermiation. Zn trafficking across the membrane is allowed by specific families of transporters known as the ZnTs, which are involved in effluent release, and the Zips, which mediate uptake. We enrolled 10 normozoospermic healthy participants in an infertility survey programme, as well as 5 patients affected by testicular germ cell cancer, and 18 patients presenting with obstructive azoospermia, without mutations of the CFTR gene, and undergoing assisted reproductive technologies. The research study was performed at our University Clinic. Semen samples, or biopsies or fine needle aspirates from the testis or epididymis, were obtained from each of the participants. Protein expression of main members of the ZnT and Zip families of Zn transporters was examined in human testis and epididymis samples by immunofluorescence. Quantification of sperm Zn content was performed by flow cytometry, atomic absorption spectrometry (AA) and autometallography. Intratubular cells of the germ line displayed a high redundancy of Zip family members involved in Zn uptake, while ZnT transporters were more represented in epididymis. Testicular and epididymal spermatozoa contained less Zn than ejaculated spermatozoa (2.56 ± 0.51 and 12.58 ± 3.16 versus 40.48 ± 12.71 ng Zn/10(6)cells, respectively). Gain of hypermotility and acrosomal reaction were significantly linked to the loss of Zn content in ejaculated spermatozoa. This was an ancillary study performed on a small cohort of normozoospermic subjects. Although these results clarify the Zn trafficking during different phases of sperm life, no conclusive information can be drawn about the fertilizing potential of sperm, and the overall pregnancy outcomes, after Zn supplementation. Our data disclose the dynamics of Zn trafficking during over the sperm lifespan. No external funding was sought or obtained for this study. No conflict of interest is declared.
Product Description
Supelco
Hydroquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Propidium iodide solution
USP
Hydroquinone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N