Merck
  • The protein phosphatase Siw14 controls caffeine-induced nuclear localization and phosphorylation of Gln3 via the type 2A protein phosphatases Pph21 and Pph22 in Saccharomyces cerevisiae.
The protein phosphatase Siw14 controls caffeine-induced nuclear localization and phosphorylation of Gln3 via the type 2A protein phosphatases Pph21 and Pph22 in Saccharomyces cerevisiae.
Journal of biochemistry (2014-10-15)
Minori Numamoto, Yu Sasano, Masataka Hirasaki, Minetaka Sugiyama, Hiromi Maekawa, Satoshi Harashima
ABSTRACT
The Saccharomyces cerevisiae Siw14, a tyrosine phosphatase involved in the response to caffeine, participates in regulation of the phosphorylation and intracellular localization of Gln3, a GATA transcriptional activator of nitrogen catabolite repression-sensitive genes. In Δsiw14 cells, the phosphorylation level of Gln3 is decreased and the nuclear localization of Gln3 is stimulated by caffeine. However, the mechanism by which Siw14 controls the localization and function of Gln3 remains unclear, although the nuclear localization of Gln3 is known to be induced by activation of the type 2A phosphatases (PP2As) Pph21 and Pph22, and the type 2A-related phosphatase Sit4. In this study, we show that the increased nuclear localization of Gln3 in response to caffeine caused by disruption of the SIW14 gene is dependent on the Sit4 and PP2A phosphatases. We also show that decreased phosphorylation of Gln3 caused by disruption of the SIW14 gene is completely suppressed by deletion of both PPH21 and PPH22, but only partially suppressed by deletion of SIT4. Taking these results together, we conclude that Siw14 functions upstream of Pph21 and Pph22 as an inhibitor of the phosphorylation and localization of Gln3, and that Sit4 acts independently of Siw14.
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Hydrobromic acid, reagent grade, 48%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Hydrogen bromide solution, 33 wt. % in acetic acid
Sigma-Aldrich
Hydrobromic acid solution, 33 wt. % in acetic acid
Sigma-Aldrich
Hydrobromic acid, ACS reagent, 48%
Sigma-Aldrich
Hydrobromic acid, 48 wt. % in H2O, ≥99.99%
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
Sigma-Aldrich
Glycerol, FCC, FG
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material