Merck
  • Long-term effect of depot medroxyprogesterone acetate on vaginal microbiota, epithelial thickness and HIV target cells.
Long-term effect of depot medroxyprogesterone acetate on vaginal microbiota, epithelial thickness and HIV target cells.
The Journal of infectious diseases (2014-03-22)
Caroline M Mitchell, Leslie McLemore, Katharine Westerberg, Rena Astronomo, Kimberly Smythe, Carolyn Gardella, Matthias Mack, Amalia Magaret, Dorothy Patton, Kathy Agnew, M Juliana McElrath, Florian Hladik, David Eschenbach
ABSTRACT
Depot medroxyprogesterone acetate (DMPA) has been linked to human immunodeficiency virus type 1 (HIV-1) acquisition. Vaginal microbiota of women using DMPA for up to 2 years were cultured. Mucosal immune cell populations were measured by immunohistological staining. Over 12 months, the proportion with H2O2-positive lactobacilli decreased (n = 32; 53% vs 27%; P = .03). Median vaginal CD3(+) cells also decreased (n = 15; 355 vs 237 cells/mm(2); P = .03), as did CD3(+)CCR5(+) cells (195 vs 128 cells/mm(2); P = .04), HLA-DR(+) cells (130 vs 96 cells/mm(2); P = .27), and HLA-DR(+)CCR5(+) cells (18 vs 10 cells/mm(2); P = .33). DMPA contraception does not increase vaginal mucosal CCR5(+) HIV target cells but does decrease CD3(+) T lymphocytes and vaginal H2O2-producing lactobacilli.
Product Description
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C