Merck
  • Low-density lipoprotein and other predictors of response with telaprevir-based therapy in treatment-experienced HCV genotype 1 patients: REALIZE study.
Low-density lipoprotein and other predictors of response with telaprevir-based therapy in treatment-experienced HCV genotype 1 patients: REALIZE study.
Liver international : official journal of the International Association for the Study of the Liver (2014-10-17)
Thomas Berg, Pietro Andreone, Stanislas Pol, Stuart Roberts, Zobair Younossi, Moises Diago, Eric J Lawitz, Roberto Focaccia, Graham R Foster, Andrzej Horban, Isabelle Lonjon-Domanec, Ralph DeMasi, Gaston Picchio, Donghan Luo, Sandra De Meyer, Stefan Zeuzem
ABSTRACT
Predictors of response to treatment with peginterferon plus ribavirin are well established. In these post-hoc analyses of the REALIZE study, we sought to identify predictors of response for telaprevir-based triple therapy. Patients from the REALIZE study with baseline data for all predictors evaluated (including baseline disease characteristics and demographics, prior treatment response and baseline laboratory assessments) were included in the post-hoc analyses (n = 465). Univariate and multivariate analyses were used to evaluate factors predicting treatment outcomes. Sustained viral response (SVR) rates were 86% in prior relapsers, 63% in prior partial responders and 32% in prior null-responders. In the final multivariate analysis, baseline factors predicting SVR were prior response to treatment [Odds ratio (OR) = 2.80; 95% confidence interval (CI), 2.13-3.69], low-density lipoprotein (LDL) (≥2.6 mmol/L) (OR = 2.11; 95% CI, 1.52-2.93), HCV genotype (OR = 0.58; 95% CI, 0.36-0.93), and maximum alanine amino transferase and aspartate amino transferase (OR = 0.62; 95% CI, 0.40-0.97). Prior response to peginterferon plus ribavirin treatment and LDL levels are the main independent predictive markers of response with telaprevir-based triple therapy.
Product Description
Ribavirin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ribavirin, antiviral
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications