Merck
  • Development of a liquid chromatography-mass spectrometry method for the determination of the neurotoxic quinolinic acid in human serum.
Development of a liquid chromatography-mass spectrometry method for the determination of the neurotoxic quinolinic acid in human serum.
Clinica chimica acta; international journal of clinical chemistry (2014-06-25)
Andreas Meinitzer, Andreas Tomaschitz, Stefan Pilz, Manfred Truber, Gabriele Zechner, Martin Gaksch, Barbara Prietl, Gerlies Treiber, Michaela Schwarz, Andreas Baranyi
ABSTRACT
Quinolinic acid (QA) is thought to be one of the most important metabolites of the kynurenine pathway with the highest biological activity in apoptotic responses and neurodegenerative diseases. The determination of QA might be of clinical relevance in different patient groups, but currently, only a few laborious methods with high levels of sample volume consumption are available. We developed and validated a simple liquid chromatography-tandem mass spectrometric (LC-tandem MS) method for the determination of QA in human serum with low sample volume requirements. The presented method provides high sample throughput with 25 μL aliquots and works in the positive electrospray ionization (ESI) mode. A commercially available QA-d3 was used as internal standard. Specific transitions for QA and QA-d3 were m/z 280→m/z 78 and m/z 283→m/z 81, respectively. The intra- and inter-assay coefficients of variation (CVs) were all below 10%. Applying this method, in 50 healthy humans a mean serum concentration of QA of 350±167 nmol/L (mean±SD) was determined. The described method is suitable for large clinical trials, which is of potential clinical importance to elucidate the function of QA and its relationship to different disease patterns and may be applicable for clinical laboratory routine.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium tetraborate, 99.998% trace metals basis
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Sodium tetraborate, 99%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
2,3-Pyridinedicarboxylic acid, 99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes