Merck
  • Effect of renal impairment on the pharmacokinetics, efficacy, and safety of albiglutide.
Effect of renal impairment on the pharmacokinetics, efficacy, and safety of albiglutide.
Postgraduate medicine (2014-06-12)
Malcolm A Young, Jeffrey A Wald, Jessica E Matthews, Fred Yang, Rickey R Reinhardt
ABSTRACT
Chronic kidney disease is frequently present in patients with type 2 diabetes mellitus (T2DM). New therapeutic options in this patient subpopulation are needed. Assess the effect of renal impairment on the pharmacokinetics (PK), efficacy, and safety of albiglutide in single- and multiple-dose studies. Pharmacokinetics, safety, and efficacy of once weekly albiglutide in patients with T2DM was assessed from a single-dose (30 mg), nonrandomized, open-label study (N = 41) including subjects with normal and varying degrees of renal impairment, including hemodialysis, and a pooled analysis of 4 phase 3, randomized, double-blind (1 open-label), active or placebo-controlled multiple-dose studies. The pooled analysis of the latter 4 studies (N = 1113) was part of the population PK analysis, which included subjects with normal and varying degrees of renal impairment (mild, moderate, severe) treated with albiglutide (30 to 50 mg) to primary end points of 26 to 52 weeks. Single-dose PK showed area-under-the-curve ratios (and 90% CIs) of 1.32 (0.96-1.80), 1.39 (1.03-1.89), and 0.99 (0.63-1.57) for the moderate, severe, and hemodialysis groups, respectively, relative to the normal group. Results indicate that modest increases in plasma concentration of albiglutide were observed with the severity of renal impairment. There was a trend for more glycemic lowering as the estimated glomerular filtration rate decreased. The severe group had a higher frequency of gastrointestinal (eg, diarrhea, constipation, nausea, and vomiting) and hypoglycemic (with background sulfonylurea use) events compared with patients with mild or moderate renal impairment. The PK, efficacy, and safety data indicate that albiglutide has a favorable benefit/risk ratio in patients with T2DM and varying degrees of renal impairment, and the need for a dose adjustment is not suggested. Experience in patients with more severe renal impairment is very limited, so the recommendation is to use albiglutide carefully in this population. (ClinicalTrials.gov):NCT00938158, NCT00849017, NCT00838916, NCT00839527, NCT0198539.
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)