Merck
  • Monitoring and preparation of neoagaro- and agaro-oligosaccharide products by high performance anion exchange chromatography systems.
Monitoring and preparation of neoagaro- and agaro-oligosaccharide products by high performance anion exchange chromatography systems.
Carbohydrate polymers (2015-03-31)
Bartosz Kazłowski, Chorng Liang Pan, Yuan Tih Ko
ABSTRACT
A series of neoagaro-oligosaccharides (NAOS) were prepared by β-agarase digestion and agaro-oligosaccharides (AOS) by HCl hydrolysis from agarose with defined quantity and degree of polymerization (DP). Chain-length distribution in the crude product mixtures were monitored by two high performance anion exchange chromatography systems coupled with a pulsed amperometric detector. Method 1 utilized two separation columns: a CarboPac(™) PA1 and a CarboPac(™) PA100 connected in series and method 2 used the PA100 alone. Method 1 resolved the product in size ranges consisting of DP 1-46 for NAOS and DP 1-32 for AOS. Method 2 clearly resolved saccharide product sizes within DP 26. The optimized system utilizing a semi-preparative CarboPac(™) PA100 column was connected with a fraction collector to isolate and quantify individually separated products. This study established systems for the preparation and qualitative and quantitative measurements as well as for the isolation of various sizes of oligomers generated from agarose.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioXtra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioReagent, suitable for electrophoresis, 98.0-102.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Galactose, European Pharmacopoeia (EP) Reference Standard
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, ACS reagent, ≥98%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, puriss. p.a., ACS reagent, ≥99.0% (T)
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis