Merck
  • Defining key signaling nodes and therapeutic biomarkers in NF1-mutant cancers.
Defining key signaling nodes and therapeutic biomarkers in NF1-mutant cancers.
Cancer discovery (2014-06-11)
Clare F Malone, Jody A Fromm, Ophélia Maertens, Thomas DeRaedt, Rachel Ingraham, Karen Cichowski
ABSTRACT
NF1 encodes a RAS GTPase-activating protein. Accordingly, aberrant RAS activation underlies the pathogenesis of NF1-mutant cancers. Nevertheless, it is unclear which RAS pathway components represent optimal therapeutic targets. Here, we identify mTORC1 as the key PI3K effector in NF1-mutant nervous system malignancies and conversely show that mTORC2 and AKT are dispensable. However, we find that tumor regression requires sustained inhibition of both mTORC1 and MEK. Transcriptional profiling studies were therefore used to establish a signature of effective mTORC1-MEK inhibition in vivo. We unexpectedly found that the glucose transporter GLUT1 was potently suppressed, but only when both pathways were inhibited. Moreover, unlike VHL- and LKB1-mutant cancers, reduction of (18)F-FDG uptake required the suppression of both mTORC1 and MEK. Together, these studies identify optimal and suboptimal therapeutic targets in NF1-mutant malignancies and define a noninvasive means of measuring combined mTORC1-MEK inhibition in vivo, which can be readily incorporated into clinical trials. This work demonstrates that mTORC1 and MEK are key therapeutic targets in NF1-mutant cancers and establishes a noninvasive biomarker of effective, combined target inhibition that can be evaluated in clinical trials.
Sigma-Aldrich
MISSION® esiRNA, targeting human MTOR
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Frap1
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution