Merck
  • Enhancement of enteric adenovirus cultivation in a novel Ras-overexpressing cell line.
Enhancement of enteric adenovirus cultivation in a novel Ras-overexpressing cell line.
The Journal of general virology (2013-10-08)
Jiyeon Si, Misoon Kim, Mi Young Lim, GwangPyo Ko
ABSTRACT
Enteric human adenoviruses (HAdVs; serotypes 40 and 41) have been identified as an emerging cause of drinking water contamination. Due to their fastidious characteristics, HAdVs are difficult to cultivate and therefore not detected easily by standard mammalian cell cultivation methods. Here we found that human embryonic kidney 293 cells, transformed transiently with Ras, enhanced HAdV replication by more than threefold. We also constructed a stable cell line overexpressing the Ras protein, 293-Ras, in which the replication of three HAdV strains of serotypes 40 and 41 was increased markedly. However, only HAdV replication was enhanced; infection of 293 and 293-Ras cells with human rhinivorus-6 showed no significant differences in replication rate. Infected 293-Ras cells exhibited an increased level and phosphorylation of extracellular regulated kinase (ERK). In addition, the Ras-mediated increase in HAdV replication was impaired by the mitogen-activated protein kinase/ERK kinase (MEK1) inhibitor U0126, suggesting direct involvement of the MEK1/ERK pathway in enhanced HAdV replication. Based on these results, we suggest that the 293-Ras cell line be used for the efficient detection and cultivation of HAdV strains in both clinical and environmental specimens.
Product Description
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC