Merck
  • Aspirated bile: a major host trigger modulating respiratory pathogen colonisation in cystic fibrosis patients.
Aspirated bile: a major host trigger modulating respiratory pathogen colonisation in cystic fibrosis patients.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (2014-05-13)
F J Reen, D F Woods, M J Mooij, M N Chróinín, D Mullane, L Zhou, J Quille, D Fitzpatrick, J D Glennon, G P McGlacken, C Adams, F O'Gara
ABSTRACT
Chronic respiratory infections are a leading global cause of morbidity and mortality. However, the molecular triggers that cause respiratory pathogens to adopt persistent and often untreatable lifestyles during infection remain largely uncharacterised. Recently, bile aspiration caused by gastro-oesophageal reflux (GOR) has emerged as a significant complication associated with respiratory disease, and cystic fibrosis (CF) in particular. Based on our previous finding that the physiological concentrations of bile influence respiratory pathogens towards a chronic lifestyle in vitro, we investigated the impact of bile aspiration on the lung microbiome of respiratory patients. Sputum samples (n = 25) obtained from a cohort of paediatric CF patients were profiled for the presence of bile acids using high-resolution liquid chromatography-mass spectrometry (LC-MS). Pyrosequencing was performed on a set of ten DNA samples that were isolated from bile aspirating (n = 5) and non-bile aspirating (n = 5) patients. Both denaturing gradient gel electrophoresis (DGGE) and pyrosequencing revealed significantly reduced biodiversity and richness in the sputum samples from bile aspirating patients when compared with non-aspirating patients. Families and genera associated with the pervasive CF microbiome dominated aspirating patients, while bacteria associated with the healthy lung were most abundant in non-aspirating patients. Bile aspiration linked to GOR is emerging as a major host trigger of chronic bacterial infections. The markedly reduced biodiversity and increased colonisation by dominant proteobacterial CF-associated pathogens observed in the sputum of bile aspirating patients suggest that bile may play a major role in disease progression in CF and other respiratory diseases.
Product Description
Lithocholic acid, European Pharmacopoeia (EP) Reference Standard
Ursodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Ursodeoxycholic acid, ≥99%
Sigma-Aldrich
Lithocholic acid, ≥95%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Chenodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chenodeoxycholic acid
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.