Merck
  • A novel combined micellar system of lapatinib and Paclitaxel with enhanced antineoplastic effect against human epidermal growth factor receptor-2 positive breast tumor in vitro.
A novel combined micellar system of lapatinib and Paclitaxel with enhanced antineoplastic effect against human epidermal growth factor receptor-2 positive breast tumor in vitro.
Journal of pharmaceutical sciences (2014-11-26)
Yan Wei, Shuai Xu, Feng Wang, Aifeng Zou, Shuang Zhang, Yan Xiong, Shilei Cao, Qizhi Zhang, Yajie Wang, Xinguo Jiang
ABSTRACT
Lapatinib (LPT) could sensitize human epidermal growth factor receptor-2 (HER-2) positive breast cancer to paclitaxel (PTX) and induce synergetic action with PTX in preclinical test and phase II/III trial. In this study, LPT-conjugated poly (ethylene glycol) (PEG) and poly (lactic acid) (PLA) (LPT-PEG-PLA) was first synthesized and confirmed with ¹H Nuclear Magnetic Resonance and Matrix-Assisted Laser Desorption/ Ionization Time of Flight Mass Spectrometry, which was used for the preparation of a novel PEG-PLA combined micelles of LPT and PTX (PPM-LP). The obtained PPM-LP exhibited uniform, spherical shape with a size of 25.80 ± 0.47 nm and zeta potential of -3.17 ± 0.15 mv. PTX existed in molecular or amorphous forms in the micelles and superficial LPT could better delay PTX release. The cytotoxicity of PPM-LP with LPT conjugation against SKBr-3 cells (HER-2 positive) was found to be significantly increasing as compared with PPM-PTX, whereas there was no significant difference against MDA-MB-231 cells (HER-2 negative). PPM-LP could escape from endosomes and be distributed into cytoplasm and led to cell arrest in G2/M and G1/S phases simultaneously. Results of nucleus staining and flow cytometry confirmed that LPT could remarkably increase antineoplastic effect of PTX against SKBr-3 cells. All these results demonstrated that PPM-LP may be a promising drug delivery system for HER-2 positive breast cancer.
Product Description
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, ≥98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%