Merck
  • Insulin like growth factor binding protein 7 (IGFBP7) expression is linked to poor prognosis but may protect from bone disease in multiple myeloma.
Insulin like growth factor binding protein 7 (IGFBP7) expression is linked to poor prognosis but may protect from bone disease in multiple myeloma.
Journal of hematology & oncology (2015-04-19)
Arnold Bolomsky, Dirk Hose, Martin Schreder, Anja Seckinger, Susanne Lipp, Bernard Klein, Daniel Heintel, Heinz Ludwig, Niklas Zojer
ABSTRACT
Insulin like growth factor binding protein 7 (IGFBP7) is a secreted protein binding insulin like growth factor 1 (IGF-1), insulin, vascular endothelial growth factor A (VEGFA), and activin A. It antagonizes bone morphogenetic proteins and is involved in the tumour propagation of solid as well as haematological malignancies. Its role in multiple myeloma (MM) is not defined so far. We therefore aim here to investigate its prognostic and pathophysiological role in MM. The clinical significance of IGFBP7 gene expression was investigated by gene expression profiling in two independent cohorts (n = 948) of newly-diagnosed MM patients. Methylation of the IGFBP7 promoter was analysed by pyrosequencing and treatment of MM cell lines with 5-aza-2-deoxycytidine. The impact of IGFBP7 on MM cells was studied by CCK-8 assay, BrdU assay and flow cytometry, respectively. IGFBP7 expression in bone marrow stromal cells (BMSCs) was studied by quantitative RT-PCR. For osteoblast development, immortalized and primary human BMSCs were cultured in osteogenic differentiation medium for 7-14 days in the presence of recombinant human IGFBP7 and/or activin A. Median IGFBP7 expression is significantly lower in CD138-purified plasma cells from individuals with MGUS and MM, compared to normal bone marrow plasma cells. IGFBP7 gene expression in MM cells is regulated by methylation, shown by pyrosequencing and exposure to demethylating agents (5-aza-2-deoxycytidine). High expression of IGFBP7 in MM cells is associated with adverse survival in two independent cohorts of 247 and 701 newly-diagnosed MM patients treated with high-dose therapy and autologous stem cell transplantation. IGFBP7 is associated with prognostically adverse chromosomal aberrations (t(4;14) and gain of 1q21), MMSET expression, and higher myeloma cell proliferation. In vitro, IGFBP7 overcomes activin A induced osteoblast suppression and promotes osteogenesis. MM cells downregulate IGFBP7 in stromal cells, possibly contributing to the osteoblast suppression found in MM. Conversely, higher IGFBP7 expression is associated with a lower probability of myeloma bone disease. Our data indicate that IGFBP7 expression is a marker for a specific methylation pattern in myeloma, linked to translocation t(4;14) associated MMSET expression, showing clinical features of adverse prognosis with absence of myeloma bone disease.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture