Merck
  • Ultrastructural morphologic changes in mycobacterial biofilm in different extreme condition.
Ultrastructural morphologic changes in mycobacterial biofilm in different extreme condition.
Ultrastructural pathology (2014-09-06)
Virendra Kumar, Tarun Kumar Sachan, Pragya Sharma, Krishna Dutta Rawat
ABSTRACT
The aim of this study was to investigate the morphologic and ultrastructural features of biofilms of slow and fast-growing mycobacteria in different stress conditions, presence and absence of oleic acid albumin dextrose catalase (OADC) enrichment and at different temperatures: 30, 37 and 42 °C. Four hundred mycobacterial isolates were taken. The biomass of each biofilm was quantified using a modified microtiter plate assay method. Isolates were divided into those that formed fully established biofilms, moderately attached biofilms and weakly adherent biofilms by comparison with a known biofilm-forming strain. The large quantity of biofilm was produced by Mycobacterium smegmatis at temperature 37 and 42 °C as compared to 30 °C. Mycobacterium fortuitum and M. avium developed large amount of biofilm at 30 °C as compared to 37 and 42 °C. Mycobacterium tuberculosis developed strong biofilm at 37 °C and no biofilm at 30 and 42 °C in Sauton's media. The selected non-tuberculous mycobacteria and H37Rv developed strong biofilm in the presence of OADC enrichment in Sauton's medium. Microscopic examination of biofilms by scanning electron microscopy revealed that poorly adherent biofilm formers failed to colonize the entire surface of the microtiter well. While moderately adherent biofilm formers grew in uniform monolayers but failed to develop a mature three-dimensional structure. SEM analysis of an isolate representative of the group formed fully established biofilms with a textured, multi-layered, three-dimensional structure.
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%