Merck
  • Gastrokine 1 inhibits the carcinogenic potentials of Helicobacter pylori CagA.
Gastrokine 1 inhibits the carcinogenic potentials of Helicobacter pylori CagA.
Carcinogenesis (2014-09-23)
Jung Hwan Yoon, Ho Suk Seo, Sung Sook Choi, Hyun Suk Chae, Won Seok Choi, Olga Kim, Hassan Ashktorab, Duane T Smoot, Suk Woo Nam, Jung Young Lee, Won Sang Park
ABSTRACT
Helicobacter pylori CagA directly injected by the bacterium into epithelial cells via a type IV secretion system, leads to cellular changes such as morphology, apoptosis, proliferation and cell motility, and stimulates gastric carcinogenesis. We investigated the effects of cytotoxin-associated gene A (CagA) and gastrokine 1 (GKN1) on cell proliferation, apoptosis, reactive oxygen species (ROS) production, epithelial-mesenchymal transition (EMT) and cell migration in CagA- or GKN1-transfected gastric epithelial cells and mucosal tissues from humans and mice infected with H.pylori. On the molecular level, H.pylori CagA induced increased cell proliferation, ROS production, antiapoptotic activity, cell migration and invasion. Moreover, CagA induced activation of NF-κB and PI3K/Akt signaling pathways and EMT-related proteins. In addition, H.pylori CagA reduced GKN1 gene copy number and expression in gastric cells and mucosal tissues of humans and mice. However, GKN1 overexpression successfully suppressed the carcinogenic effects of CagA through binding to CagA. These results suggest that GKN1 might be a target to inhibit the effects from H.pylori CagA.
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
4-Hydroxy-TEMPO, 95%
Sigma-Aldrich
Propidium iodide solution
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium fluoride, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
SAFC
Sodium deoxycholate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%