Merck
  • The metabolism of lysophosphatidic acids by allelic variants of human soluble epoxide hydrolase.
The metabolism of lysophosphatidic acids by allelic variants of human soluble epoxide hydrolase.
Drug metabolism and pharmacokinetics (2015-03-12)
Endang R Purba, Elsa A Leuhery, Ami Oguro, Susumu Imaoka
ABSTRACT
Lysophosphatidic acids (LPAs) are phospholipids which have many physiological and pathophysiological functions. The human soluble epoxide hydrolase (sEH) plays a role in the metabolism of xenobiotics through its metabolism of aromatic hydrocarbon epoxides such as styrene oxide. sEH also has a phosphatase activity, and metabolizes LPAs. In this study, we investigated a purified wild-type (WT) and six allelic variants of sEH to evaluate differences in their activities toward LPAs. We found that the R103C and R287Q showed significantly lower activity than the WT sEH. We also found that the R103C and R287Q had significantly lower activity even when applied to only the N-terminal or C-terminal domain. The kinetic study determined that the R103C and R287Q had a lower Vmax/Km ratio toward stearoyl-LPA than the other variants. In a previous study, we found that WT sEH suppressed vascular endothelial growth factor (VEGF) mRNA in Hep3B cells; in the present experiments, all sEH variants except V442A suppressed VEGF mRNA levels in Hep3B cells. These results suggest that the R103C and R287Q have lower phosphatase activity, but that all the allelic variants have similar effects on VEGF suppression.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
4-Methylumbelliferyl phosphate
Sigma-Aldrich
4-Methylumbelliferyl phosphate disodium salt
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
trans-Stilbene oxide, 98%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Geranylgeranyl pyrophosphate ammonium salt, solution, ≥95% (TLC), ~1 mg/mL in methanol: NH4OH (7:3)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard