Merck
  • A high dose of short term exogenous D-galactose administration in young male rats produces symptoms simulating the natural aging process.
A high dose of short term exogenous D-galactose administration in young male rats produces symptoms simulating the natural aging process.
Life sciences (2015-02-01)
Saida Haider, Laraib Liaquat, Sidrah Shahzad, Sadia Sadir, Syeda Madiha, Zehra Batool, Saiqa Tabassum, Sadia Saleem, Fizza Naqvi, Tahira Perveen
ABSTRACT
D-Galactose (D-gal) induced accelerated senescence has been used to develop an aging model for brain. Previously, long term administration of a wide range of doses has been used for this purpose. In the present study we investigate whether short term administration of a high dose of D-gal in rats induces significant signs and symptoms similar to natural aging. Young rats were injected intraperitoneally with D-gal at a dose of 300 mg/ml/kg for one week. Behavioral analysis for depression and anxiety like symptoms were monitored by forced swim test (FST) and light/dark transition (LDT) test. Assessment of memory was done using the Morris water maze (MWM), passive avoidance test (PAT) and elevated plus maze (EPM) test. Biochemical analysis was done for estimation of antioxidant enzymes and acetylcholinesterase. Determination of brain biogenic amines was performed by HPLC-EC. Short term administration of D-gal significantly altered behavioral, biochemical and neurochemical responses in rats. D-Gal injected rats exhibited depressogenic and anxiogenic behaviors while memory was also significantly impaired in these rats. Brain lipid peroxidation and superoxide dismutase activity were significantly increased while catalase and glutathione peroxidase decreased. Increased activity of acetylcholinesterase was also exhibited by D-gal injected rats while brain biogenic amines were significantly decreased. Food intake and growth rate were however comparable in both groups. Together the behavioral, biochemical and neurochemical impairments following the high dose of D-gal suggest that symptoms similar to natural aging may be developed in rats in as early as one week.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydroxylamine hydrochloride, ACS reagent, 98.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydroxylamine hydrochloride, ReagentPlus®, 99%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydroxylamine hydrochloride, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N