Merck
  • Effects of progesterone and norethindrone on female fathead minnow (Pimephales promelas) steroidogenesis.
Effects of progesterone and norethindrone on female fathead minnow (Pimephales promelas) steroidogenesis.
Environmental toxicology and chemistry (2014-12-04)
Lene H Petersen, David Hala, Dennis Carty, Mark Cantu, Dalma Martinović, Duane B Huggett
ABSTRACT
As knowledge of contaminants capable of adversely modulating endocrine functions increases, attention is focused on the effects of synthetic progestins as environmental endocrine disrupters. In the present study, effects of exposure to a synthetic progestin (norethindrone, 168 ± 7.5 ng/L) and endogenous progestogen (progesterone, 34 ± 4.1 ng/L) on steroidogenesis in adult female fathead minnows were examined. In vivo exposure to either compound lowered expression (nonsignificant) of luteinizing hormone (LHβ) levels in the brain along with significantly down-regulating the beta isoform of membrane progesterone receptor (mPRβ) in ovary tissue. The correspondence between lowered LHβ levels in the brain and mPRβ in the ovary is suggestive of a possible functional association as positive correlations between LHβ and mPR levels have been demonstrated in other fish species. In vitro exposure of ovary tissue to progesterone resulted in significantly elevated progestogen (pregnenolone, 17α-hydroxyprogesterone, and 17α,20β-dihydroxypregnenone) and androgen (testosterone) production. Whereas in vitro exposure to norethindrone did not significantly impact steroid hormone production but showed decreased testosterone production relative to solvent control (however this was not significant). Overall, this study showed that exposure to a natural progestogen (progesterone) and synthetic progestin (norethindrone), was capable of modulating LHβ (in brain) and mPRβ expression (in ovary).
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Supelco
Ethyl acetate, analytical standard