Merck
  • A coumarin-based fluorescent probe as a central nervous system disease biomarker.
A coumarin-based fluorescent probe as a central nervous system disease biomarker.
Sensors (Basel, Switzerland) (2014-11-13)
Ann-Chee Yap, Ummi Affah Mahamad, Shen-Yang Lim, Hae-Jo Kim, Yeun-Mun Choo
ABSTRACT
Homocysteine and methylmalonic acid are important biomarkers for diseases associated with an impaired central nervous system (CNS). A new chemoassay utilizing coumarin-based fluorescent probe 1 to detect the levels of homocysteine is successfully implemented using Parkinson's disease (PD) patients' blood serum. In addition, a rapid identification of homocysteine and methylmalonic acid levels in blood serum of PD patients was also performed using the liquid chromatography-mass spectrometry (LC-MS). The results obtained from both analyses were in agreement. The new chemoassay utilizing coumarin-based fluorescent probe 1 offers a cost- and time-effective method to identify the biomarkers in CNS patients.
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
DL-Homocysteine, ≥95% (titration)
Sigma-Aldrich
Methylmalonic acid, 99%
Supelco
DL-Homocysteine, analytical standard