Merck
  • Hispidin derived from Phellinus linteus affords protection against acrylamide-induced oxidative stress in Caco-2 cells.
Hispidin derived from Phellinus linteus affords protection against acrylamide-induced oxidative stress in Caco-2 cells.
Chemico-biological interactions (2014-06-01)
Wei Chen, Yang Shen, Hongming Su, Xiaodong Zheng
ABSTRACT
Acrylamide (AA), a well-known toxicant, has attracted numerous attentions for its presumably carcinogenesis, neurotoxicity and cytotoxicity. Oxidative stress was considered to be associated with acrylamide cytotoxicity, but the link between oxidative stress and acrylamide cytotoxicity is still unclear. In the present study, hispidin produced from the edible fungus Phellinus linteus displayed dramatically antioxidant activities against DPPH radicals, ABTS radicals, ferric reducing and hydroxyl radicals, as well as superoxide anion radicals. Moreover, the cytoprotective effect of hispidin against AA-induced oxidative stress was verified upon Caco-2 cells according to evaluate the cell viability, intracellular ROS, mitochondrial membrane potential (MMP) and glutathione (GSH) in the presence or absence of AA (5 mM) in a dose-dependent manner. Collectively, our results demonstrated for the first time that hispidin was able to inhibit AA-induced oxidative stress, which might have implication for the dietary preventive application.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Decylamine, 95%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Rhodamine 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Decylamine, ≥99.0% (GC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Sigma-Aldrich
2,3-Naphthalenedicarboxaldehyde, suitable for fluorescence