Merck
  • Glycyrol suppresses collagen-induced arthritis by regulating autoimmune and inflammatory responses.
Glycyrol suppresses collagen-induced arthritis by regulating autoimmune and inflammatory responses.
PloS one (2014-07-19)
Yanxia Fu, Hailing Zhou, Shuangyan Wang, Qun Wei
ABSTRACT
Glycyrol is a natural compound extracted from Glycyrrhiza uralensis, first reported by us to be a new immunosuppressant. Here, we demonstrate its beneficial effect in collagen-induced arthritis (CIA) in mice, a model for rheumatoid arthritis (RA) in man, and we document the underlying mechanisms. Peroral administration of glycyrol significantly reduced clinical scores, alleviated cartilage and bone erosion and reduced levels of serum inflammatory cytokines. Glycyrol also decreased delayed-type hypersensitivity, improved carbon clearance and reduced acetic acid-induced capillary permeability. Furthermore, glycyrol decreased NF-κB and NFAT transcriptional activities and inhibited IL-2 expression. The therapeutic effect of glycyrol was associated with down-regulation of both autoimmune and inflammatory reactions. In addition, we demonstrated that glycyrol has minimal acute toxicity in mice. Therefore, we propose that glycyrol may hold promise for future treatment of RA.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)