Merck
  • Isolation of Escherichia coli strains with AcrAB-TolC efflux pump-associated intermediate interpretation or resistance to fluoroquinolone, chloramphenicol and aminopenicillin from dogs admitted to a university veterinary hospital.
Isolation of Escherichia coli strains with AcrAB-TolC efflux pump-associated intermediate interpretation or resistance to fluoroquinolone, chloramphenicol and aminopenicillin from dogs admitted to a university veterinary hospital.
The Journal of veterinary medical science (2014-03-22)
Toyotaka Sato, Shin-ichi Yokota, Risa Ichihashi, Tomoka Miyauchi, Torahiko Okubo, Masaru Usui, Nobuhiro Fujii, Yutaka Tamura
ABSTRACT
Understanding the prevalence of antimicrobial-resistance and the relationship between emergence of resistant bacteria and clinical treatment can facilitate design of effective treatment strategies. We here examined antimicrobial susceptibilities of Escherichia coli isolated from dogs admitted to a university hospital (University hospital) and companion animal clinics (Community clinics) in the same city and investigated underlying multidrug-resistance mechanisms. The prevalence of E. coli with intermediate and resistant interpretations to ampicillin (AMP), enrofloxacin (ENR) and chloramphenicol (CHL) was higher in the University hospital than in the Community clinics cases. Use of antimicrobials, including fluoroquinolone, was also significantly higher in the University hospital than in the Community clinics cases. Upon isolation using ENR-supplemented agar plates, all ENR-resistant isolates had 3-4 nucleotide mutations that accompanied by amino acid substitutions in the quinolone-resistance-determining regions of gyrA, parC and parE, and 94.7% of all isolates derived from the University hospital showed AMP and/or CHL resistance and possessed blaTEM and/or catA1. The average mRNA expression levels of acrA, acrB and tolC and the prevalence of organic solvent tolerance, in isolates derived from ENR-supplemented agar plates were significantly higher in the University hospital than in the Community clinics isolates. Thus, E. coli derived from the University hospital cases more often showed concomitant decreased susceptibilities to aminopenicillins, fluoroquinolones and CHL than did those derived from the Community clinics; this was related to an active AcrAB-TolC efflux pump, in addition to acquisition of specific resistance genes and genetic mutations.
Product Description
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Supelco
Enrofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Enrofloxacin, United States Pharmacopeia (USP) Reference Standard
Enrofloxacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
USP
Cephalexin, United States Pharmacopeia (USP) Reference Standard
Supelco
Cefalexin, VETRANAL®, analytical standard
Millipore
Oxytetra Selective Supplement, suitable for microbiology
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
Ampicillin
Cefazolin, European Pharmacopoeia (EP) Reference Standard
USP
Cefazolin, United States Pharmacopeia (USP) Reference Standard
Supelco
Oxytetracycline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ampicillin, meets USP testing specifications