Merck
  • Regulatory T cell activity is partly inhibited in a mouse model of chronic Pseudomonas aeruginosa lung infection.
Regulatory T cell activity is partly inhibited in a mouse model of chronic Pseudomonas aeruginosa lung infection.
Experimental lung research (2014-11-15)
Feng-Ming Ding, Song-Lei Zhu, Ce Shen, Xiao-Li Ji, Xin Zhou
ABSTRACT
We aimed to investigate the activity of regulatory T (Treg) cells in chronic Pseudomonas aeruginosa (PA) lung infection and its influence on effector T-cell responses. C57BL/6 mice were randomly inoculated with PA-laden agarose beads (1 × 10(5) CFU/50 μL) or planktonic PA (1 × 10(5) CFU/50 μL), and euthanized at the time points of 4 hour, day 1, 3, 5, and 7. Bacterial load, bronchoalveolar lavage (BAL) fluid cell counts, and lung tissue histology were assessed. BAL fluid concentrations of TGF-β1, IFN-γ, IL-1β, IL-4, IL-6, IL-10, and IL-17A were measured. Messenger RNA (mRNA) levels of TGF-β1, IL-10 and CD4(+) T-cell subtype-specific transcription factors were determined. The expression of CD4(+)CD25(+)forkhead box P3 (FoxP3)(+) cells in lungs and spleens were analyzed. Mice inoculated with PA-laden agarose beads developed chronic PA lung infection during 7-day study period, while mice inoculated with planktonic PA cleared bacteria in 3 days. Compared with mice recovered from acute PA lung infection, those with chronic infection had significantly increased effector T-cell responses, accompanied by a more severe neutrophilic inflammation. Mice with chronic PA lung infection had significantly lower concentration of TGF-β1 and higher concentrations of IFN-γ, IL-1β, IL-6, and IL-17A in BAL fluid. Meanwhile, they had significantly lower mRNA levels of TGF-β1, IL-10 and FoxP3 in lung tissues, and lower expression of CD4(+)CD25(+)FoxP3(+) cells in lungs and spleens. These findings indicate that Treg cell activity is partly inhibited in mice with chronic PA lung infection, which contributes to the enhanced effector T-cell responses in airways.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
3,4,4′-Trichlorocarbanilide, 99%
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)