Merck
  • Flavonoid profile and antioxidant activities of methanolic extract of Hyparrhenia hirta (L.) Stapf.
Flavonoid profile and antioxidant activities of methanolic extract of Hyparrhenia hirta (L.) Stapf.
Indian journal of experimental biology (2015-05-28)
Hanen Bouaziz-Ketata, Nabil Zouari, Hichem Ben Salah, Moez Rafrafi, Najiba Zeghal
ABSTRACT
In this study, we report isolation of flavonoids, viz., 3-O-methylquercetin, tangeritin, luteolin-7-O-glucoside, luteolin, apigenin-7-O-glucoside, apigenin-8-C-glucoside, luteolin-8-C-glucoside, luteolin-6-C-glucoside, diosmetin and catechin from the methanolic extract of Hyparrhenia hirta employing high performance liquid chromatography and liquid chromatography-electrospray ionization-tandem mass spectrometry. The total phenolic content of H. hirta extract was 105.58 ± 0.1 mg gallic acid equivalents/g of plant extract while the total flavonoid content was 45.20 ± 0.2 mg quercetin equivalents/g of plant extract and the total condensed tannin were 72.35 ± 0.7 mg catechin equivalents/g of plant extract by reference to standard curve. The antioxidant activity was assayed through the antioxidant capacity by phosphomolybdenum assay, the reducing power assay and the radical scavenging activity using 2,2-diphenyl-1-picrylhydrazyl method. The extract showed dose dependant activity in all the three assays.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Aluminum chloride, anhydrous, sublimed, ≥98%
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
USP
Vanillin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%