Merck
  • Saturation solubility of nicotine, scopolamine and paracetamol in model stratum corneum lipid matrices.
Saturation solubility of nicotine, scopolamine and paracetamol in model stratum corneum lipid matrices.
International journal of pharmaceutics (2014-07-06)
Anne Mundstock, Geoffrey Lee
ABSTRACT
The saturation solubilities of nicotine and scopolamine bases as well as acidic paracetamol were measured in two different model stratum corneum lipid matrices. Light microscopy visualized the presence of drug above its solubility either as droplets or crystalline particles. Neither wide-angle X-ray diffraction nor DSC detected the drugs. The saturation solubilities of the nicotine and scopolamine bases are 3-5% w/w and 5-10% w/w respectively. Paracetamol strongly disrupts the lamellar phase formed by the lipids and could be dissolved to >20% w/w. Based on these results the saturation solubilities of nicotine and scopolamine in an intact stratum corneum membrane are estimated to be up to 17.5 μg and 35 μg drug per milligrams of stratum corneum membrane respectively. These concentrations are well above values obtained by tape stripping of intact stratum corneum during a permeation experiment. The drug's saturation solubility in the stratum corneum's lipid phase is therefore unlikely to be rate-limiting for permeation of these two drugs.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder