Merck
  • P63 alpha mutations lead to aberrant splicing of keratinocyte growth factor receptor in the Hay-Wells syndrome.
P63 alpha mutations lead to aberrant splicing of keratinocyte growth factor receptor in the Hay-Wells syndrome.
The Journal of biological chemistry (2003-04-15)
Alexey Fomenkov, Yi-Ping Huang, Ozlem Topaloglu, Anna Brechman, Motonobo Osada, Tanya Fomenkova, Eugene Yuriditsky, Barry Trink, David Sidransky, Edward Ratovitski
ABSTRACT
p63, a p53 family member, is required for craniofacial and limb development as well as proper skin differentiation. However, p63 mutations associated with the ankyloblepharon-ectodermal dysplasia-clefting (AEC) syndrome (Hay-Wells syndrome) were found in the p63 carboxyl-terminal region with a sterile alpha-motif. By two-hybrid screen we identified several proteins that interact with the p63alpha carboxyl terminus and its sterile alpha-motif, including the apobec-1-binding protein-1 (ABBP1). AEC-associated mutations completely abolished the physical interaction between ABBP1 and p63alpha. Moreover the physical association of p63alpha and ABBP1 led to a specific shift of FGFR-2 alternative splicing toward the K-SAM isoform essential for epithelial differentiation. We thus propose that a p63alpha-ABBP1 complex differentially regulates FGFR-2 expression by supporting alternative splicing of the K-SAM isoform of FGFR-2. The inability of mutated p63alpha to support this splicing likely leads to the inhibition of epithelial differentiation and, in turn, accounts for the AEC phenotype.