Merck
  • Low RPS14 expression is common in myelodysplastic syndromes without 5q- aberration and defines a subgroup of patients with prolonged survival.
Low RPS14 expression is common in myelodysplastic syndromes without 5q- aberration and defines a subgroup of patients with prolonged survival.
Haematologica (2009-10-02)
Akos Czibere, Ingmar Bruns, Bärbel Junge, Raminder Singh, Guido Kobbe, Rainer Haas, Ulrich Germing
ABSTRACT
To further clarify the role of ribosomal protein S14 (RPS14) in myelodysplastic syndrome, we examined RPS14 transcription in bone marrow derived CD34+ cells from patients with non-5q- myelodysplastic syndrome and found a reduced expression of RPS14 in 51 of 72 (71%) patients. MDS patients with an intermediate-1 risk (INT-1) score according to the international prognostic scoring system and low RPS14 expression had a superior median overall survival of not reached versus 25 months compared to INT-1 patients with high RPS14 expression (p=0.0249). Using multivariate analysis, the RPS14 expression status was confirmed as an independent predictor for survival in INT-1 patients.