Merck
  • Forming End-to-End Oligomers of Gold Nanorods Using Porphyrins and Phthalocyanines.
Forming End-to-End Oligomers of Gold Nanorods Using Porphyrins and Phthalocyanines.
Langmuir : the ACS journal of surfaces and colloids (2015-06-09)
Alexander F Stewart, Brandon P Gagnon, Gilbert C Walker
ABSTRACT
The illumination of aggregated metal nanospecies can create strong local electric fields to brighten Raman scattering. This study describes a procedure to self-assemble gold nanorods (NRs) through the use of porphyrin and phthalocyanine agents to create reproducibly stable and robust NR aggregates in the form of end-to-end oligomers. Narrow inter-rod gaps result, creating electric field "hot spots" between the NRs. The organic linker molecules themselves are potential Raman-based optical labels, and the result is significant numbers of Raman-active species located in the hot spots. NR polymerization was quenched by phospholipid encapsulation, which allows for control of the polydispersity of the aggregate solution, to optimize the surface-enhanced Raman scattering (SERS) enhancement and permitted the aqueous solubility of the aggregates. The increased presence of Raman-active species in the hot spots and the optimizing of solution polydispersity resulted in the observation of scattering enhancements by encapsulated porphyrins/phthalocyanines of up to 3500-fold over molecular chromophores lacking the NR oligomer host.
Sigma-Aldrich
Copper(II) phthalocyanine, sublimed grade, Dye content 99 %
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Copper(II) phthalocyanine, Dye content >99 %
Sigma-Aldrich
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Copper(II) phthalocyanine, β-form, Dye content 90 %
Sigma-Aldrich
Copper(II) phthalocyanine, triple-sublimed grade, >99.95% trace metals basis
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes