Merck
  • Quantitative hopanoid analysis enables robust pattern detection and comparison between laboratories.
Quantitative hopanoid analysis enables robust pattern detection and comparison between laboratories.
Geobiology (2015-04-14)
C-H Wu, L Kong, M Bialecka-Fornal, S Park, A L Thompson, G Kulkarni, S J Conway, D K Newman
ABSTRACT
Hopanoids are steroid-like lipids from the isoprenoid family that are produced primarily by bacteria. Hopanes, molecular fossils of hopanoids, offer the potential to provide insight into environmental transitions on the early Earth, if their sources and biological functions can be constrained. Semiquantitative methods for mass spectrometric analysis of hopanoids from cultures and environmental samples have been developed in the last two decades. However, the structural diversity of hopanoids, and possible variability in their ionization efficiencies on different instruments, have thus far precluded robust quantification and hindered comparison of results between laboratories. These ionization inconsistencies give rise to the need to calibrate individual instruments with purified hopanoids to reliably quantify hopanoids. Here, we present new approaches to obtain both purified and synthetic quantification standards. We optimized 2-methylhopanoid production in Rhodopseudomonas palustris TIE-1 and purified 2Me-diplopterol, 2Me-bacteriohopanetetrol (2Me-BHT), and their unmethylated species (diplopterol and BHT). We found that 2-methylation decreases the signal intensity of diplopterol between 2 and 34% depending on the instrument used to detect it, but decreases the BHT signal less than 5%. In addition, 2Me-diplopterol produces 10× higher ion counts than equivalent quantities of 2Me-BHT. Similar deviations were also observed using a flame ionization detector for signal quantification in GC. In LC-MS, however, 2Me-BHT produces 11× higher ion counts than 2Me-diplopterol but only 1.2× higher ion counts than the sterol standard pregnane acetate. To further improve quantification, we synthesized tetradeuterated (D4) diplopterol, a precursor for a variety of hopanoids. LC-MS analysis on a mixture of (D4)-diplopterol and phospholipids showed that under the influence of co-eluted phospholipids, the D4-diplopterol internal standard quantifies diplopterol more accurately than external diplopterol standards. These new quantitative approaches permit meaningful comparisons between studies, allowing more accurate hopanoid pattern detection in both laboratory and environmental samples.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
Squalene, ≥98%, liquid
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
1,3-Propanediol, 98%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Disodium succinate, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)