Merck
  • Light-modulated seminal wavy roots in rice mediated by nitric oxide-dependent signaling.
Light-modulated seminal wavy roots in rice mediated by nitric oxide-dependent signaling.
Protoplasma (2015-01-27)
Hsiang-Wen Chen, Ko-Hsuan Shao, Shu-Jen Wang
ABSTRACT
Rice (Oryza sativa L.) seminal roots from germinated seeds help establish seedlings, but the seminal root growth and morphology are sensitive to environmental factors. Our previous research showed that several indica-type rice varieties such as Taichung native 1 (TCN1) showed light-induced wavy roots. Also, auxin and oxylipins are two signaling factors regulating the wavy root photomorphology. To investigate the signaling pathway, here, we found that nitric oxide (NO) was a second messenger triggering the signal transduction of light stimuli to induce the wavy morphology of seminal roots in rice. Moreover, interactions between oxylipins and phytohormones such as ethylene and auxin participating in the NO-dependent regulatory pathway of light-induced wavy roots were examined. The order of action of signaling components in the pathway was NO, oxylipins, ethylene, and auxin.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
1-Naphthylacetic acid, 1 mg/mL, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ethylene, ≥99.5%
Sigma-Aldrich
1-Naphthaleneacetic acid, technical grade
Sigma-Aldrich
2-(p-Chlorophenoxy)-2-methylpropionic acid, 97%
Sigma-Aldrich
1-Naphthaleneacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
2-Aminoisobutyric acid, 98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC