Merck
  • High-level expression and characterization of a thermophilic β-mannanase from Aspergillus niger in Pichia pastoris.
High-level expression and characterization of a thermophilic β-mannanase from Aspergillus niger in Pichia pastoris.
Biotechnology letters (2015-05-15)
Shi Yu, Zhezhe Li, Yaping Wang, Wanping Chen, Lin Fu, Wei Tang, Chen Chen, Yunyun Liu, Xue Zhang, Lixin Ma
ABSTRACT
A novel, high-level expression, thermostable mannan endo-1,4-beta-mannosidase is urgently needed for industrial applications. The mannan endo-1,4-β-mannosidase gene (MAN) from Aspergillus niger CBS 513.88 was optimized based on the codon usage bias in Pichia pastoris and synthesized by overlapping PCR to produce MAN-P. It was expressed in P. pastoris GS115 from a constitutive expression vector pHBM-905 M. MAN-P reached 594 mg/l in shake-flasks after 192 h induction. On production in a 5 l fermenter, the yield of MAN-P reached ~3.5 mg/ml and the enzyme activity was 1612 U/ml. The enzyme exhibited a maximum activity of 3049 U/ml at 80 °C and retained 60% enzyme activity at 80 °C for 2 h. The pH optimum was 4.5 and the enzyme was stable over the pH range 1.5-11. The thermostability of MAN-P is higher than other known fungal mannanases and the expression and thermophilic properties make MAN-P useful for industrial applications.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)