Merck
  • Effect of osteopontin on the mRNA expression of ADAMTS4 and ADAMTS5 in chondrocytes from patients with knee osteoarthritis.
Effect of osteopontin on the mRNA expression of ADAMTS4 and ADAMTS5 in chondrocytes from patients with knee osteoarthritis.
Experimental and therapeutic medicine (2015-07-03)
Shu-Guang Gao, Chao Zeng, Yang Song, Jian Tian, Chao Cheng, Tuo Yang, Hui Li, Fang-Jie Zhang, Guang-Hua Lei
ABSTRACT
Previous studies have demonstrated that osteopontin (OPN) levels are elevated in the synovial fluid and articular cartilage, and are associated with the severity of knee osteoarthritis (OA). However, the role of OPN in the pathogenesis of OA has yet to be elucidated. The present study aimed to investigate the effects of OPN on the expression of the aggrecanases, a disintegrin and metalloproteinase with thrombospondin motifs (ADAMTS)4 and ADAMTS5, in human OA chondrocytes, as they serve a key function in aggrecan degradation. Human OA chondrocytes were obtained from the knees of 16 patients with OA, and subsequently cultured in a monolayer. The chondrocytes were divided into three groups, which included the control (no treatment), N-OPN (treated with 100 ng/ml OPN, the normal circulating OPN concentration) and the H-OPN groups (treated with 1 µg/ml OPN, a high OPN concentration). Reverse transcription-quantitative polymerase chain reaction was performed to quantify the relative mRNA expression levels of ADAMTS4, ADAMTS5 and aggrecan in the chondrocytes. The mRNA expression levels of ADAMTS4 were significantly reduced in the N-OPN and H-OPN groups when compared with the control group (P<0.0001). In addition, the mRNA expression levels of ADAMTS4 were lower in the H-OPN group when compared with the N-OPN group (P<0.001). However, no statistically significant difference was observed in the relative mRNA expression levels of ADAMTS5 among the three groups (P>0.05). Furthermore, the mRNA expression levels of aggrecan were higher in the N-OPN and H-OPN groups when compared with the control group (P<0.0001), and a statistically significant difference was observed between the N-OPN and H-OPN groups with regard to the mRNA expression of aggrecan (P<0.0001). These results demonstrated that OPN may exert a protective effect in human OA chondrocytes against aggrecan degradation by suppressing the expression of ADAMTS4.
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%