Merck
  • Evaluation of small noncoding RNAs in ex vivo stored human mature red blood cells: changes in noncoding RNA levels correlate with storage lesion events.
Evaluation of small noncoding RNAs in ex vivo stored human mature red blood cells: changes in noncoding RNA levels correlate with storage lesion events.
Transfusion (2015-07-16)
Tewarit Sarachana, Sandhya Kulkarni, Chintamani D Atreya
ABSTRACT
While biomarkers of storage lesions (SLs) for red blood cells (RBCs) abound, the physiologic consequences of SLs and associated important events are poorly understood. Previously we have identified differentially expressed regulatory small noncoding RNAs (ncRNAs) in stored RBCs, suggesting their role in the RBC SL process and their potential as quality biomarkers of stored RBCs. Comprehensive ncRNA expression analysis of RBCs stored for up to 56 days was performed on RNAs collected from enriched mature RBCs on Days 0, 7, 14, 28, 42, and 56. Three known RBC SL processes, that is, mature RBCs' suicidal death (eryptosis), ATP loss, and changes in RBC indices, were correlated with differentially expressed ncRNAs to gain knowledge on the SL molecular processes. The analysis identified four ncRNAs whose changes in the expression levels were correlated with the selected three SL processes. Differential expression on Days 14 and 28 of the four selected ncRNAs was confirmed by TaqMan quantitative reverse transcription-polymerase chain reaction analysis. Bioinformatics analysis identified potential targets and biologic functions of these ncRNAs. Overexpression of one such ncRNA, hsa-miR-196a, in a human erythroblast cell line confirmed its protective effects against the cell death and ATP loss. Overall, this study demonstrates that changes in the levels of small ncRNAs of stored RBCs correlate with some of the SL events and thus they have the potential to serve as the storage quality markers.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%