Merck
  • TRIF adaptor signaling is important in abdominal aortic aneurysm formation.
TRIF adaptor signaling is important in abdominal aortic aneurysm formation.
Atherosclerosis (2015-06-24)
Emina Vorkapic, Anna M Lundberg, Mikko I Mäyränpää, Per Eriksson, Dick Wågsäter
ABSTRACT
Abdominal aortic aneurysm (AAA) is characterized by inflammation, loss of smooth muscle cells (SMCs), and degradation of the extracellular matrix in the vessel wall. Innate immune receptors such as Toll-like receptors (TLRs) were recently shown to regulate immunological processes leading to the formation and progression of atherosclerotic plaques as well as to other cardiovascular pathologies. Our aim was to investigate whether blockage of TLR signaling, under the control of TIR domain-containing adaptor protein including IFN-β (TRIF), could inhibit the inflammatory response and AAA development in mice. In human AAA, an increased TLR3 and TLR4 expression in association with macrophages and T lymphocytes was demonstrated with immunohistochemical analysis. Angiotensin (Ang) II-induced aneurysm formation was significantly reduced by 30% in ApoE(-/-)Trif(-/-) mice compared to ApoE(-/-) mice. Morphologically, AngII-infused ApoE(-/-)Trif(-/-) mice had a more intact cellular and extracellular matrix while ApoE(-/-) mice infused with AngII displayed an increased medial thickness associated with aortic dissection, thrombus formation, and a more disorganized vessel wall. Gene expression analysis of the abdominal aorta revealed a profound decrease of the inflammatory genes CD68 (P < 0.05), CD11b (P < 0.05), and TNF-α (P < 0.05) and the protease gene MMP-12 (P < 0.01) in ApoE(-/-)Trif(-/-) mice compared to ApoE(-/-) mice infused with AngII. Our results suggest that signaling through TRIF is important for the inflammatory response of AngII-induced AAA and that blockage of the TRIF pathway reduces vascular inflammation and protects against AAA formation.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications