Merck
  • Metallosupramolecular Ni2L3 and Ni4L6 complexes of bis-bidentate pyridine-containing ligands: X-ray structures and catalytic proton reduction.
Metallosupramolecular Ni2L3 and Ni4L6 complexes of bis-bidentate pyridine-containing ligands: X-ray structures and catalytic proton reduction.
Dalton transactions (Cambridge, England : 2003) (2015-06-25)
Chui-Shan Tsang, Lingjing Chen, Lu-Wei Li, Shek-Man Yiu, Tai-Chu Lau, Hoi-Lun Kwong
ABSTRACT
The pyridine-containing ligands and react with Ni(2+) ions to afford dinuclear triple-stranded helicate and tetrahedral cage supramolecular complexes, respectively; the two architectures exhibit contrasting reactivity towards electro- and photocatalytic proton reduction.
Sigma-Aldrich
2-Pyridinecarboxaldehyde, 99%
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)