Merck
  • A novel human STAT3 mutation presents with autoimmunity involving Th17 hyperactivation.
A novel human STAT3 mutation presents with autoimmunity involving Th17 hyperactivation.
Oncotarget (2015-09-08)
Judith Wienke, Willemijn Janssen, Rianne Scholman, Hilde Spits, Marielle van Gijn, Marianne Boes, Joris van Montfrans, Nicolette Moes, Sytze de Roock
ABSTRACT
Mutations in STAT3 have recently been shown to cause autoimmune diseases through increased lymphoproliferation. We describe a novel Pro471Arg STAT3 mutation in a patient with multiple autoimmune diseases, causing hyperactivation of the Th17 pathway. We show that IL-17 production by primary T cells was enhanced and could not be further increased by IL-6, while IL-10 reduced Th17 cell numbers. Moreover, specific inhibition of STAT3 activation resulted in diminished IL-17 production. We show that the Pro471Arg STAT3 mutation yields both increased levels of IgA and IgG, probably due to high IL-21 levels. When remission was reached through medical intervention, IL-17 levels normalized and the clinical symptoms improved, supporting the idea that STAT3 gain-of-function mutations can cause hyperactivation of the Th17 pathway and thereby contribute to autoimmunity.
Sigma-Aldrich
IL-6 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Interleukin-6 from rat, IL-6, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-6 from mouse, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, carrier free
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture