Merck
  • Fabrication of novel magnetic nanocomposite with a number of adsorption sites for the removal of dye.
Fabrication of novel magnetic nanocomposite with a number of adsorption sites for the removal of dye.
International journal of biological macromolecules (2015-03-24)
Leilei Li, Huimin Duan, Xiaojiao Wang, Chuannan Luo
ABSTRACT
In this current work, a novel functional ionic liquid-coated Fe3O4@chitosan@graphene oxide was synthesized and tested as adsorbents for methylene blue through batch adsorption tests. The physicochemical properties of the adsorbent were investigated using scanning electron microscopy, x-ray diffraction, and fourier transform infrared spectroscopy. The adsorption capacity of the adsorbent was evaluated as a function of initial dye concentration, pH, contact time and temperature. The adsorption results indicated that the adsorption followed a pseudo-second-order kinetics. The equilibrium adsorption was well-described by the Langmuir isotherm model and the maximum adsorption capacity was to be 262 mg/g. The negative value of ΔH° shows exothermic nature of adsorption process and the negative value of ΔG° indicated that the adsorption reaction is spontaneous. Moreover, the ionic liquid-coated Fe3O4@chitosan@graphene oxide could be repeatedly used by simple treatment without obvious structure and performance degradation. The obtained results suggested that the potential applications of ionic liquid-coated Fe3O4@chitosan@graphene oxide in deep-purification of polluted water.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Tetraoctylammonium bromide, 98%