Merck
  • Analysis of gait in rats with olivocerebellar lesions and ability of the nicotinic acetylcholine receptor agonist varenicline to attenuate impairments.
Analysis of gait in rats with olivocerebellar lesions and ability of the nicotinic acetylcholine receptor agonist varenicline to attenuate impairments.
Behavioural brain research (2015-06-07)
C S Lambert, R M Philpot, M E Engberg, B E Johns, L Wecker
ABSTRACT
Studies have demonstrated that administration of the neuronal nicotinic receptor agonist varenicline to rats with olivocerebellar lesions attenuates balance deficits on a rotorod and balance beam, but the effects of this drug on gait deficits have not been investigated. To accomplish this, male Sprague-Dawley rats were trained to walk on a motorized treadmill at 25 and 35 cm/s and baseline performance determined; both temporal and spatial gait parameters were analyzed. A principal component analysis (PCA) was used to identify the key components of gait, and the cumulative gait index (CGI) was calculated, representing deviations from prototypical gait patterns. Subsequently, animals either remained as non-lesioned controls or received injections of 3-acetylpyridine (3-AP)/nicotinamide to destroy the climbing fibers innervating Purkinje cells. The gait of the non-lesioned group was assessed weekly to monitor changes in the normal population, while the gait of the lesioned group was assessed 1 week following 3-AP administration, and weekly following the daily administration of saline or varenicline (0.3, 1.0, or 3.0mg free base/kg) for 2 weeks. Non-lesioned animals exhibited a 60-70% increased CGI over time due to increases in temporal gait measures, whereas lesioned animals exhibited a nearly 3-fold increased CGI as a consequence of increases in spatial measures. Following 2 weeks of treatment with the highest dose of varenicline (3.0mg free base/kg), the swing duration of lesioned animals normalized, and stride duration, stride length and step angle in this population did not differ from the non-lesioned population. Thus, varenicline enabled animals to compensate for their impairments and rectify the timing of the gait cycle.
Sigma-Aldrich
Varenicline tartrate, ≥98% (HPLC)
Sigma-Aldrich
3-Aminopyrazole, 98%
Sigma-Aldrich
3-AP, ≥98% (HPLC)