Merck
Production and Characterization of New Anti-HER2 Monoclonal Antibodies.
Monoclonal antibodies in immunodiagnosis and immunotherapy (2015-06-20)
Manijeh Mahdavi, Mehrnaz Keyhanfar, Abbas Jafarian, Hassan Mohabatkar, Mohammad Rabbani
ABSTRACT
Breast cancer is a major public health problem worldwide. Although in Iran cancer is the third cause of death after coronary heart disease and accidents, mortality from cancer has been on the rise during recent decades. About 15% to 20% of patients with invasive breast cancer have abnormally high levels of HER2 protein. HER2 is a specialized protein found on breast cancer cells that controls cancer growth and spread. This study describes the generation and characterization of new anti-HER2 MAbs towards HER2 protein using a chimeric peptide immunogen containing discontinuous B-cell epitope peptide (peptide 626) and promiscuous T-helper epitope (MVF). The specificity of these MAbs was confirmed in various immunoassays, including ELISA, Western blotting, and immunofluorescence. Moreover, the MTT assay results indicated that 5H5 and 5H11 MAbs could reduce the growth of SKBR3 cells by approximately 50% (p<0.05). These MAbs that can reduce cancer cell proliferation would be useful for cancer therapy. Furthermore, the synthetic peptide used in the current work was able to induce the immune system to generate antibodies, especially IgG isotype. Therefore, it could be further used as a peptide cancer vaccine that targets different epitopes or structural domains of HER2 ECD.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture