Merck
  • One-step production of biodiesel from oils with high acid value by activated Mg-Al hydrotalcite nanoparticles.
One-step production of biodiesel from oils with high acid value by activated Mg-Al hydrotalcite nanoparticles.
Bioresource technology (2015-06-29)
Yi-Tong Wang, Zhen Fang, Fan Zhang, Bao-Jin Xue
ABSTRACT
Activated Mg-Al hydrotalcite (HT-Ca) nanoparticles (<45 nm) were synthesized by co-precipitation and hydrothermal activation with aqueous Ca(OH)2 solution. They were characterized by various techniques including X-ray diffraction, inductively coupled plasma atomic-emission spectrometer, Brunauer-Emmett-Teller method, scanning electronic microscope-X-ray energy dispersive analysis and temperature programmed desorption method. HT-Ca presented both acidic and basic due to the formation of Mg4Al2(OH)14 · 3H2O, Mg2Al(OH)7 and AlO(OH) nanocrystals to esterify and transesterify oils with high acid value (AV). Under conditions of 5 wt% HT-Ca, 160 °C, 30/1 methanol/oil molar ratio and 4h, 93.4% Jatropha biodiesel yield was obtained at AV of 6.3 mg KOH/g with 4 cycles (biodiesel yield>86%). It was further found that it can resist free fatty acids, and biodiesel yield reached 92.9% from soybean oil with high AV of 12.1. HT-Ca catalyst showed a potential practical application for direct production of biodiesel from oils with high AV without pretreatment.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Heptadecanoic acid, ≥98%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O