Merck
  • Preliminary characterization and expression of Vasa-like gene in Schistosoma japonicum.
Preliminary characterization and expression of Vasa-like gene in Schistosoma japonicum.
Parasitology research (2015-04-23)
Yujie Diao, Mengqing Hua, Yanjing Shao, Wei Huang, Miao Liu, Cuiping Ren, Yongsheng Ji, Jianmin Chen, Jijia Shen
ABSTRACT
The Vasa gene is a vital germline marker to study the origin and development of germ cells and gonads in many organisms. Until now, little information was available about the characteristics of the Vasa gene in Schistosoma japonicum (S. japonicum). In this study, we cloned the open reading frame (ORF) of the S. japonicum Vasa-like gene (Sj-Vasa). The expression pattern and tissue localization of Sj-Vasa were also analyzed. Our results showed that Sj-Vasa shared the general feature of DEAD-box family member proteins. Sj-Vasa was transcribed and expressed throughout the S. japonicum life cycle with transcription exhibiting high levels at day 24 in both male and female worms, and the expression level in the female was always higher than that in the male. Sj-Vasa protein was localized in a variety of tissues of adult schistosomes, including the gonads (ovary, vitellarium, and testes), the subtegument, and some cells of the parenchyma. To our knowledge, this is the first report of preliminary characterization and expression of the Vasa-like gene that may play an important role in the development of the worm, especially in reproductive organs of S. japonicum.
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
SAFC
Isopropyl β-D-1-thiogalactopyranoside